Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VTFALHSAS VFCVSLDPKT NTLAVTGGED DKAFVWRLSD GELLFECAGH KDSVTCAGFS HDSTLVATGD MSGLLKVWQV DTKEEVWSFE AGDLEWMEWH PRAPVLLAGT ADGNTWMWKV PNGDCKTFQG PNCPATCGRV LPDGKRAVVG YEDGTIRIWD LKQGSPIHVL KGTEGHQGPL TCVAANQDGS LILTGSVDCQ AKLVSATTGK VVGVFRPETV ASQPSLGEGE ESESNSVESL GFCSVMPLAA VGYLDGTLAI YDLATQTLRH QCQHQSGIVQ LLWEAGTAVV YTCSLDGIVR LWDARTGRLL TDYRGHTAEI LDFALSKDAS LVVTTSGDHK AKVFCVQRPD R (85-434 aa encoded by BC039866)
The gene product is an immunoglobulin-type protein. It is found to be expressed strongly in endothelial cells, cytotrophoblasts, and poorly differentiated colon adenocarcinoma cells found in lymphatics. The protein contains a heparin-binding domain and mediates heparin-sensitive cell adhesion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Angio-associated migratory cell protein; angio-associated migratory protein; angio-associated, migratory cell protein
Gene Aliases: AAMP; Aamp-rs; AU040907
UniProt ID: (Human) Q13685
Entrez Gene ID: (Human) 14, (Rat) 301512, (Mouse) 227290
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support