Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This gene encodes a member of the acyl-CoA dehydrogenase family. Members of this family of proteins localize to the mitochondria and catalyze the rate-limiting step in the beta-oxidation of fatty acyl-CoA. The encoded protein is specifically active toward palmitoyl-CoA and long-chain unsaturated substrates. Mutations in this gene cause acyl-CoA dehydrogenase family member type 9 deficiency. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACAD-9; Acyl-CoA dehydrogenase family member 9; acyl-CoA dehydrogenase family member 9, mitochondrial; acyl-CoA dehydrogenase family, member 9; acyl-Coenzyme A dehydrogenase family, member 9; Complex I assembly factor ACAD9, mitochondrial; very-long-chain acyl-CoA dehydrogenase VLCAD; very-long-chain acyl-CoA dehydrogenase VLCAD homolog
Gene Aliases: 2600017P15Rik; 4732402K02; ACAD9; C630012L17Rik; NPD002; Nyggf2
UniProt ID: (Human) Q9H845, (Rat) B1WC61, (Mouse) Q8JZN5
Entrez Gene ID: (Human) 28976, (Rat) 294973, (Mouse) 229211
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support