Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TLLYRHGDR SPVKTYPKDP YQEEEWPQGF GQLTKEGMLQ HWELGQALRQ RYHGFLNTSY HRQEVYVRST DFDRTLMSAE ANLAGLFPPN GMQRFNPNIS WQPIPVHTVP ITEDRLLKFP LGPCPRYEQL QNETRQTPEY QNESSRNAQF LDMVANETGL TDLTLETVWN VYDTLFCEQT HGLRLPPWAS PQTMQRLSRL KDFSFRFLFG IYQQAEKARL QGGVLLAQIR KNLTLMATTS QLPKLLVYSA HDTTLVALQM ALDVYNGEQA PYASCHIFEL YQEDSGNFSV EMYFRNESDK APWPLSLPGC PHRCPLQDFL RLTEPVVPKD WQQECQLASG PADTEVIV (37-383 aa encoded by BC003160 )
This gene encodes the beta subunit of lysosomal acid phosphatase (LAP). LAP is chemically and genetically distinct from red cell acid phosphatase. The encoded protein belongs to a family of distinct isoenzymes which hydrolyze orthophosphoric monoesters to alcohol and phosphate. Mutations in this gene or in the related alpha subunit gene cause acid phosphatase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acid phosphatase 2, lysozymal; LAP; LMW-PTP-II low molecular weight phosphotyrosine protein phosphatase isozyme 2; Lysosomal acid phosphatase
Gene Aliases: Acp-2; ACP2; LAP
UniProt ID: (Human) P11117, (Mouse) P24638
Entrez Gene ID: (Human) 53, (Rat) 24162, (Mouse) 11432
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support