Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: DRSVLAKEL KFVTLVFRHG DRSPIDTFPT DPIKESSWPQ GFGQLTQLGM EQHYELGEYI RKRYRKFLNE SYKHEQVYIR STDVDRTLMS AMTNLAALFP PEGVSIWNPI LLWQPIPVHT VPLSEDQLLY LPFRNCPRFQ ELESETLKSE EFQKRLHPYK DFIATLGKLS GLHGQDLFGI WSKVYDPLYC ESVHNFTLPS WATEDTMTKL RELSELSLLS LYGIHKQKEK SRLQGGVLVN EILNHMKRAT QIPSYKKLIM YSAHDTTVSG LQMALDVYNG LLPPYASCHL TELYFEKGEY FVEMYYRNET QHEPYPLMLP GCSPSCPLER FAELAGPVIP QDWSTECM (27-373 aa encoded by BC007460 )
ACPP is an enzyme which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-NT; 5'-nucleotidase; Acid phosphatase 3; EC 3.1.3.2; Ecto-5'-nucleotidase; Fluoride-resistant acid phosphatase; FRAP; PAP; PPAP; prostate acid phosphatase; Prostatic acid phosphatase; prostatic acid phosphatase (rPAP); prostatic acid phosphotase; Protein tyrosine phosphatase ACP3; Thiamine monophosphatase; TMPase
Gene Aliases: 5'-NT; ACP-3; ACP3; ACPP; Acpp11; FRAP; pap; Ppal; RNACPP11
UniProt ID: (Human) P15309, (Rat) P20646
Entrez Gene ID: (Human) 55, (Rat) 56780
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support