Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MGSQVSVESG ALHVVIVGGG FGGIAAASQL QALNVPFMLV DMKDSFHHNV AALRASVETG FAKKTFISYS VTFKDNFRQG LVVGIDLKNQ MVLLQGGEAL PFSHLILATG STGPFPGKFN EVSSQQAAIQ AYEDMVRQVQ RSRFIVVVGG GSAGVEMAAE IKTEYPEKEV TLIHSQVALA DKELLPSVRQ EVKEILLRKG VQLLLSERVS NLEELPLNEY REYIKVQTDK GTEVATNLVI LCTGIKINSS AYRKAFESRL ASSGALRVNE HLQVEGHSNV YAIGDCADVR TPKMAYLAGL HANIAVANIV NSVKQRPLQA YKPGALTFLL SMGRNDGVGQ ISGFYVGRLM VRLTKSRDLF VSTSWKTMRQ SPP
The p53 tumor suppressor protein induces cell cycle arrest or apoptosis in response to cellular stresses. PRG3 (p53-responsive gene 3), is induced specifically under p53-dependent apoptotic conditions in human colon cancer cells. PRG3 has significant homology to bacterial oxidoreductases and the apoptosis-inducing factor, AIF. Unlike AIF, PRG3 localizes in the cytoplasm and its ectopic expression induces apoptosis. The PRG3 gene is thus a novel p53 target gene in a p53-dependent apoptosis pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AMID; apoptosis-inducing factor (AIF)-homologous mitochondrion-associated inducer of death; apoptosis-inducing factor (AIF)-like mitochondrion-associated inducer of death; apoptosis-inducing factor 2; Apoptosis-inducing factor homologous mitochondrion-associated inducer of death; apoptosis-inducing factor, mitochondrion-associated, 2; apoptosis-inducing factor-like mitchondrion-associated inducer of death; apoptosis-inducing factor-like mitochondrion-associated inducer of death; Ferroptosis suppressor protein 1; FSP1; MBP2; MBPH; MGC126662; MGC141971; p53-responsive gene 3 protein; RP11-367H5.2
Gene Aliases: 5430437E11Rik; AIFM2; AMID; D730001I10Rik; PRG3
UniProt ID: (Human) Q9BRQ8, (Mouse) Q8BUE4
Entrez Gene ID: (Human) 84883, (Mouse) 71361
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support