Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GDFFSYTRHE PVGVCGQIIP WNFPLLMQAW KLGPALATGN VVVMKVAEQT PLTALYVANL IKEAGFPPGV VNIVPGFGPT AGAAIASHED VDKVAFTGST EIGRVIQVAA GSSNLKRVTL ELGGKSPNII MSDADMDWAV EQAHFALFFN QGQCCCAGSR TFVQEDIYDE FVERSVARAK SRVVGNPFDS KTEQGPQVDE TQFKKILGYI NTGKQEGAKL LCGGGIAADR GYFIQPTVFG DVQDGMTIAK EEIFGPVMQI LKFKTIEEVV GRANNSTYGL AAAVFTKDLD KANYLSQALQ AGTVWVNCYD VFGAQSPFGG YKMSGSGREL GEYGLQAYTE VKTVTVKVPQ KNS
ALDH2 belongs to the aldehyde dehydrogenase family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. Most Caucasians have two major isozymes, while approximately 50% of Asians have only the cytosolic isozyme, missing the mitochondrial isozyme. A remarkably higher frequency of acute alcohol intoxication among Asians than among Caucasians could be related to the absence of the mitochondrial isozyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetaldehyde dehydrogenase 2; AHD-M1; aldehyde dehydrogenase 2 family (mitochondrial); aldehyde dehydrogenase 2, mitochondrial; Aldehyde dehydrogenase, mitochondrial; ALDH class 2; ALDH-E2; ALDH1; ALDHI; EC 1.2.1.3; liver mitochondrial ALDH; MGC1806; mitochondrial aldehyde dehydrogenase; mitochondrial aldehyde dehydrogenase 2; nucleus-encoded mitochondrial aldehyde dehydrogenase 2
Gene Aliases: Ahd-1; Ahd-5; AHD-M1; Ahd1; Ahd5; ALDH-E2; ALDH2; ALDHI; ALDM
UniProt ID: (Human) P05091, (Pig) Q2XQV4, (Rat) P11884, (Mouse) P47738
Entrez Gene ID: (Human) 217, (Rabbit) 100338523, (Pig) 733685, (Rat) 29539, (Mouse) 11669
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support