Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total AIBP.
The product of this gene interacts with apolipoprotein A-I (apoA-I), the major apolipoprotein of high-density lipoproteins (HDLs). It is secreted into some bodily fluids, and its synthesis and secretion are stimulated in vitro by incubating cells with apoA-I. The human genome contains related pseudogenes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AI-BP; AI-BP {ECO:0000255|HAMAP-Rule:MF_03159}; apoA-I binding protein; apolipoprotein A-I binding protein; apolipoprotein A-I interacting protein; Apolipoprotein A-I-binding protein; apolipoprotein A-I-binding protein {ECO:0000255|HAMAP-Rule:MF_03159}; MGC119143; MGC119144; MGC119145; NAD(P)H-hydrate epimerase; NAD(P)H-hydrate epimerase {ECO:0000255|HAMAP-Rule:MF_03159}; NAD(P)HX epimerase; NAD(P)HX epimerase {ECO:0000255|HAMAP-Rule:MF_03159}; NAXE; YjeF N-terminal domain-containing protein 1; YjeF_N1
Gene Aliases: AA087124; AI-BP; AIBP; APOA1BP; Apoa1ip; ESTM37; NAXE; YJEFN1
UniProt ID: (Mouse) Q8K4Z3, (Human) Q8NCW5
Entrez Gene ID: (Rat) 295229, (Mouse) 246703, (Human) 128240
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support