Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: FLQAPRSPRE ERSPQEETPE GPTDAPAADE PPSELVPGPP CVANASANAT ADFEQLPARI QDFLRYRHCR HFPLLWDAPA KCAGGRGVFL LLAVKSAPEH YERRELIRRT WGQERSYGGR PVRRLFLLGT PGPEDEARAE RLAELVALEA REHGDVLQWA FADTFLNLTL K; Positive Samples: Rat stomach; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein
The protein encoded by this gene is a beta-1,3-N-acetylglucosaminyltransferase that adds an N-acetylglucosamine moiety to N-acetylgalactosamine-modified serine or threonine. The encoded enzyme is responsible for creating the core 3 structure of O-glycans, which are important components of mucin-type glycoproteins.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Gene Aliases: RGD1310926
Entrez Gene ID: (Rat) 292325
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support