Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bardet-Biedl syndrome 12 protein; Bardet-Biedl syndrome 12 protein homolog; Chaperonin-containing T-complex member BBS12; DN-452A22.3
Gene Aliases: BBS12; C4orf24; Gm1805; Gm407; Gm721
UniProt ID: (Human) Q6ZW61, (Mouse) Q5SUD9
Entrez Gene ID: (Human) 166379, (Mouse) 241950
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support