Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: EPVEAPALPA PAAPMVSKGA ESSRSSEESS SDSGSSDSEE ERATRLAELQ EQLKAVHEQL AALSQAPVNK PKKKKEKKEK EKKKKDKEKE KEKHKVKAEE EKKAKVAPPA KQAQQKKAPA KKANSTTTAG RDHFLTCGV
This gene was identified based on its homology to the gene encoding the RING3 protein, a serine/threonine kinase. The gene localizes to 9q34, a region which contains several major histocompatibility complex (MHC) genes. The function of the encoded protein is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bromodomain-containing 3; Bromodomain-containing protein 3; bromodomain-containing protein 3 short isoform; RING3-like protein
Gene Aliases: BRD3; KIAA0043; ORFX; RING3L
UniProt ID: (Human) Q15059
Entrez Gene ID: (Human) 8019
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support