Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Recommended positive controls: NT2D1, mouse testis.
Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (98%), Chicken (99%), Rhesus Monkey (100%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BTBD10 appears to behave as a suppressor of cell death, which includes neuronal cell death related to amyotrophic lateral sclerosis. It may also act as an enhancer of cell growth via its positive regulation of Akt phosphorylation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB (broad-complex tramtrack and bric-a-brac/pox virus and zinc finger) domain-containing protein 10; BTB (POZ) domain containing 10; BTB/POZ domain-containing protein 10; Glucose metabolism-related protein 1; K+ channel tetramerization protein
Gene Aliases: 1110056N09Rik; BTBD10; GMRP-1; GMRP1
UniProt ID: (Human) Q9BSF8, (Mouse) Q80X66
Entrez Gene ID: (Human) 84280, (Mouse) 68815
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support