Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAATKRKRR GGFAVQAKKP KRNEIDAEPP AKRHATAEEV EEEERDRIPG PVCKGKWKNK ERILIFSSRG INFRTRHLMQ DLRMLMPHSK ADTKMDRKDK LFVINEVCEM KNCNKCIYFE AKKKQDLYMW LSNSPHGPSA KFLVQNIHTL AELKMTGNCL KGSRPLLSFD PAFDELPHYA LLKELLIQIF STPRYHPKSQ PFVDHVFTFT ILDNRIWFRN FQIIEEDAAL VEIGPRFVLN LIKIFQGSFG GPTLYENPHY QSPNMHRRVI RSITAAKYRE KQQVKDVQKL RKKEPKTLLP HDPTADVFVT PAEEKPIEIQ WVKPEPKVDL KARKKRIYKR QRKMKQRMDS GKTK (1-353 aa encoded by BC001546)
Required for biogenesis of the 60S ribosomal subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brix domain containing 2; Brix domain-containing protein 2; BRX1, biogenesis of ribosomes, homolog; ribosome biogenesis protein Brix; Ribosome biogenesis protein BRX1 homolog
Gene Aliases: 1110064N10Rik; BRIX; BRIX1; BXDC2; C76935; RGD1308508
UniProt ID: (Human) Q8TDN6, (Mouse) Q9DCA5, (Rat) Q4QQT6
Entrez Gene ID: (Human) 55299, (Mouse) 67832, (Rat) 294799
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support