Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Positive Samples: NCI-H460, Mouse brain, Mouse thymus, Rat brain
Immunogen sequence: GALSLPSTHS CDRYPTLRSH RSSPYPSPYA HRNNSPTYSD NSPACLSMLQ SHDNWSSLGM PAHPSMLPVS HNASPPTSSS QYPSLWSVSN GAVTPGSQAA AVSNGLGAQF FRGSPAHYTP LTHPVSAPSS SGSPLYEGAA AATDIVDSQY DAAAQGRLIA SWTPVSPPSM
The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brachyury homolog; Brachyury protein; brachyury-like 2; brachyury-like 3; low ratio; Protein T; RP11-459F1.1; T brachyury homolog; T, brachyury homolog; T-box transcription factor T
Gene Aliases: Bra; cou; D17Mit170; Low; Lr; me75; SAVA; T; T1; TBXT; TFT; Tl2; Tl3
UniProt ID: (Human) O15178, (Mouse) P20293
Entrez Gene ID: (Human) 6862, (Rat) 292301, (Mouse) 20997
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support