Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTSPEIASL SWGQMKVKGS NTTYKDCKVW PGGSRTWDWR ETGTEHSPGV QPADVKEVVE KGVQTLVIGR GMSEALKVPS STVEYLKKHG IDVRVLQTEQ AVKEYNALVA QGVRVGGVFH STC (1-122 aa encoded by BC002752)
AAMDC (C11orf67) may play a role in preadipocyte differentiation and adipogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adipogenesis associated Mth938 domain-containing protein; adipogenesis associated, Mth938 domain containing; LI2; Mth938 domain-containing protein; UPF0366 protein C11orf67; UPF0366 protein C11orf67 homolog
Gene Aliases: 1810020D17Rik; 1810037D19Rik; AAMDC; C11orf67; CK067; LI2; PTD015; RGD1561459
UniProt ID: (Human) Q9H7C9, (Mouse) Q8R0P4
Entrez Gene ID: (Human) 28971, (Mouse) 66273, (Rat) 361606
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support