Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The exact function of C11orf74 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Intraflagellar transport-associated protein; Protein HEPIS
Gene Aliases: C11orf74; HEPIS; IFTAP; NWC
UniProt ID: (Human) Q86VG3
Entrez Gene ID: (Human) 119710
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support