Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
The function of C12orf45 has not been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC40397; NOP protein chaperone 1; uncharacterized protein C12orf45 homolog
Gene Aliases: AI747614; C12orf45; C430041I18Rik; D10Wsu102e; NOPCHAP1
UniProt ID: (Human) Q8N5I9, (Mouse) Q9CX66
Entrez Gene ID: (Human) 121053, (Mouse) 28109
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support