Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LINSQPKSRK TSTLQTVRIE RSPLLDQVQT FLPQMARANE KLRKEMAAAP PGRFNIENID GPHSKVIQMD VALFEMNQSD SKEVD
Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 79%.
The function of C12orf45 has not been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC40397; NOP protein chaperone 1
Gene Aliases: C12orf45; NOPCHAP1
UniProt ID: (Human) Q8N5I9
Entrez Gene ID: (Human) 121053
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support