Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPFTLHLRS RLPSAIRSLI LQKKPNIRNT SSMAGELRPA SLVVLPRSLA PAFERFCQVN TGPLPLLGQS EPEKWMLPPQ GAISET (1-85 aa encoded by BC065558)
C14orf159 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-glutamate cyclase, mitochondrial
Gene Aliases: C14orf159; DGLUCY; UNQ2439/PRO5000
UniProt ID: (Human) Q7Z3D6
Entrez Gene ID: (Human) 80017
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support