Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MLPSLQESMD GDEKELESSE EGGSAEERRL EPPSSSHYCL YSYRGSRLAQ QRGDSEDGSP SGTNAETPSG DDFSLSLADT NLPSEVEPEL RSFIAKRLSR
As component of the WDR11 complex acts together with TBC1D23 to facilitate the golgin-mediated capture of vesicles generated using AP-1 (PubMed:29426865). May have a role in spermatogenesis. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein Njmu-R1; sensitization to ricin complex subunit 2; spermatogenesis-related protein
Gene Aliases: C17orf75; NJMU-R1; SRI2
UniProt ID: (Mouse) Q9CYI0, (Human) Q7Z2H4
Entrez Gene ID: (Mouse) 70591, (Human) 64149, (Rat) 303764
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support