Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAAALRRF WSRRRAEAGD AVVAKPGVWA RLGSWARALL RDYAEACRDA SAEARARPGR AAVYVGLLGG AAACFTLAPS EGAFEEALLE ASGTLLLLAP ATRNRESEAF VQRLLWLRGR GRLRYVNLGL CSLVYEAPFD AQASLYQARC RYLQPRWTDF PGRVLDVGFV GRWWVLGAWM RDCDINDDEF LHLPAHLRVV GPQQLHSETN ERLFDEKYKP VVLTDDQVDQ ALWEEQVLQK EKKDRLALSQ AHSLVQAEAP R (1-260 aa encoded by BC005075)
C19orf52 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial import inner membrane translocase subunit Tim29; TIM29; uncharacterized protein C19orf52 homolog
Gene Aliases: c19orf52; TIMM29
UniProt ID: (Mouse) Q8BGX2, (Human) Q9BSF4
Entrez Gene ID: (Mouse) 69773, (Human) 90580
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support