Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May regulate the number of excitatory synapses that are formed on hippocampus neurons. Has no effect on inhibitory synapses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C1q and tumor necrosis factor related protein 10; C1q and tumor necrosis factor-related protein 10; C1q-domain containing protein; C1q/TNF-related protein 10; C1qTNF10; Complement C1q-like protein 2; complement component 1, q subcomponent-like 2
Gene Aliases: BC040774; C1QL2; C1QTNF10; CTRP10
UniProt ID: (Human) Q7Z5L3, (Mouse) Q8CFR0
Entrez Gene ID: (Human) 165257, (Mouse) 226359, (Rat) 288979
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support