Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total MMTAG2.
MMTAG2 gene ontology (GO) annotations include neutrophil degranulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hMMTAG2; multiple myeloma transforming 2; Multiple myeloma tumor-associated protein 2; Multiple myeloma tumor-associated protein 2 homolog
Gene Aliases: C1orf35; MMTAG2
UniProt ID: (Mouse) Q99LX5, (Human) Q9BU76
Entrez Gene ID: (Mouse) 67862, (Human) 79169
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support