Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Involved in the maturation of mitochondrial 4Fe-4S proteins functioning late in the iron-sulfur cluster assembly pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IBA57, iron-sulfur cluster assembly homolog; iron-sulfur cluster assembly factor for biotin synthase- and aconitase-like mitochondrial proteins, with a mass of 57kDa; Iron-sulfur cluster assembly factor homolog; putative transferase C1orf69 homolog, mitochondrial; putative transferase C1orf69, mitochondrial; Putative transferase CAF17 homolog, mitochondrial; Putative transferase CAF17, mitochondrial
Gene Aliases: 4930543L23Rik; A230051G13Rik; C1orf69; IBA57; MMDS3; RGD1306000; SPG74
UniProt ID: (Human) Q5T440, (Mouse) Q8CAK1
Entrez Gene ID: (Human) 200205, (Rat) 363611, (Mouse) 216792
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support