Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Immunogen sequence: TPSPQQARVC PPHMLPEDGA NLSSARGILS LIQSSTRRAY QQILDVLDEN RRPVLRGGSA AATSNPHHDN VRYGISNIDT TIEGTSDDLT VVDAASLRRQ I
Plays a role in mitochondrial and peroxisomal fission. Promotes the recruitment and association of the fission mediator dynamin-related protein 1 (DNM1L) to the mitochondrial surface. May be involved in regulation of synaptic vesicle membrane dynamics by recruitment of DNM1L to clathrin-containing vesicles. [Uniprot]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial fission factor
Gene Aliases: 5230400G24Rik; AI314724; Mff; RGD1310230
UniProt ID: (Rat) Q4KM98, (Mouse) Q6PCP5
Entrez Gene ID: (Rat) 301563, (Mouse) 75734
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support