Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
C4orf32 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: uncharacterized protein C4orf32 homolog; Uncharacterized protein FAM241A
Gene Aliases: 2700063A17Rik; C4orf32; C80506; FAM241A
UniProt ID: (Mouse) Q9CZL2, (Human) Q8N8J7
Entrez Gene ID: (Mouse) 70617, (Human) 132720, (Rat) 691931
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support