Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 93%; Dog: 93%; Horse: 86%; Human: 100%; Pig: 93%; Rat: 86%
ZC2HC1B gene ontology annotations related to this gene include metal ion binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 164, member B; protein FAM164B; Zinc finger C2HC domain-containing protein 1B; zinc finger, C2HC-type containing 1B
Gene Aliases: C6orf94; dJ468K18.5; FAM164B; ZC2HC1B
UniProt ID: (Human) Q5TFG8
Entrez Gene ID: (Human) 153918
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support