Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
C7orf72 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Post-meiotic spermatogenesis protein 1; Protein SPMIP7; Sperm microtubule inner protein 7; Spermatogenesis-associated protein 48; uncharacterized protein C7orf72 homolog
Gene Aliases: Pms1; Spata48; Spmip7
UniProt ID: (Mouse) Q5NC83
Entrez Gene ID: (Mouse) 73862, (Human) 100130988, (Rat) 289778
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support