Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKAKRSHQA IIMSTSLRVS PSIHGYHFDT ASRKKAVGNI FENTDQESLE RLFRNSGDKK AEERAKIIFA IDQDVEEKTR ALMALKKRTK DKLFQFLKLR KYSIKVH (1-106 aa encoded by BC021672)
This gene encodes a small, monomeric, predominantly unstructured protein that functions as a positive regulator of the Wnt/beta-catenin signaling pathway. This protein interacts with a repressor of beta-catenin mediated transcription at nuclear speckles. It is thought to competitively block interactions of the repressor with beta-catenin, resulting in up-regulation of beta-catenin target genes. The encoded protein may also play a role in the NF-kappaB and ERK1/2 signaling pathways. Expression of this gene may play a role in the proliferation of several types of cancer including thyroid cancer, breast cancer and hematological malignancies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: human thyroid cancer 1; TC-1; Thyroid cancer protein 1; thyroid cancer-1; Transcriptional and immune response regulator; uncharacterized protein C8orf4 homolog
Gene Aliases: 1110065B09Rik; AW121743; AW321058; C8orf4; TC-1; TC1; TCIM
UniProt ID: (Mouse) Q9D915, (Human) Q9NR00
Entrez Gene ID: (Mouse) 69068, (Human) 56892
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support