Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Specifically required during spermatogenesis for flagellum morphogenesis and sperm motility. May be required to suppress the formation of supernumerary axonemes and ensure a correct ultrastructure.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 157; uncharacterized protein C9orf117 homolog
Gene Aliases: C9orf117; CFAP157
UniProt ID: (Mouse) Q0VFX2, (Human) Q5JU67, (Rat) Q4V7B0
Entrez Gene ID: (Mouse) 227736, (Human) 286207, (Rat) 499781
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support