Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
C9orf171 is a protein coding gene. Gene ontology (GO) annotation include cilium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 77; FLJ46082; uncharacterized protein C9orf171 homolog
Gene Aliases: C9orf171; CFAP77; Gm823
UniProt ID: (Mouse) A0A087WRI3, (Human) Q6ZQR2
Entrez Gene ID: (Mouse) 329375, (Human) 389799, (Rat) 499765
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support