Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AhpC/TSA antioxidant enzyme domain-containing protein 1; Peroxiredoxin-like 2C; Thioredoxin-like protein AAED1; UPF0308 protein C9orf21; UPF0308 protein C9orf21 homolog
Gene Aliases: 1110018J18Rik; AAED1; AI849003; C9orf21; PRXL2C
UniProt ID: (Human) Q7RTV5, (Mouse) Q9D1A0
Entrez Gene ID: (Human) 195827, (Rat) 498685, (Mouse) 66129
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support