Product References
Human papillomavirus infection can alter the level of tumour stemness and T cell infiltration in patients with head and neck squamous cell carcinoma.
Frontiers in immunology
Meng L,Lu H,Li Y,Zhao J,He S,Wang Z,Shen J,Huang H,Xiao J,Sooranna SR,Song J
MA1-21454 was used in Immunohistochemistry to suggest that the stemness features of tumour cells are not only associated with the prognostic risk, but that it could also affect the immune cell interactions and associated signalling pathways.
Fri Nov 25 00:00:00 EST 2022
Long-Term Stable Mixed Chimerism after Hematopoietic Stem Cell Transplantation in Patients with Non-Malignant Disease, Shall We Be Tolerant?
PloS one
Stikvoort A,Sundin M,Uzunel M,Gertow J,Sundberg B,Schaffer M,Mattsson J,Uhlin M
MA1-21454 was used in flow cytometry to test if the two hematopoietic systems in patients with mixed chimerism remain functional and study immunological differences
Mon Jul 17 00:00:00 EDT 2017
Diversity, cellular origin and autoreactivity of antibody-secreting cell population expansions in acute systemic lupus erythematosus.
Nature immunology
Tipton CM,Fucile CF,Darce J,Chida A,Ichikawa T,Gregoretti I,Schieferl S,Hom J,Jenks S,Feldman RJ,Mehr R,Wei C,Lee FE,Cheung WC,Rosenberg AF,Sanz I
MA1-21454 was used in flow cytometry to examine the antibody repertoire of patients with acute systemic lupus erythematosus.
Wed Jul 01 00:00:00 EDT 2015
A revised strategy for monitoring BKV-specific cellular immunity in kidney transplant patients.
Kidney international
Weist BJ,Wehler P,El Ahmad L,Schmueck-Henneresse M,Millward JM,Nienen M,Neumann AU,Reinke P,Babel N
MA1-21454 was used in flow cytometry to examine the effects of immunosuppressive agents on Polyomavirus BKV-specific T cells
Tue Dec 01 00:00:00 EST 2015
Phase I Hepatic Immunotherapy for Metastases Study of Intra-Arterial Chimeric Antigen Receptor-Modified T-cell Therapy for CEA+ Liver Metastases.
Clinical cancer research : an official journal of the American Association for Cancer Research
Katz SC,Burga RA,McCormack E,Wang LJ,Mooring W,Point GR,Khare PD,Thorn M,Ma Q,Stainken BF,Assanah EO,Davies R,Espat NJ,Junghans RP
MA1-21454 was used in flow cytometry to assess intrahepatic delivery of anti-CEA chimeric antigen receptor-modified T cells through percutaneous hepatic artery infusions
Wed Jul 15 00:00:00 EDT 2015
Evidence for the involvement of gamma delta T cells in the immune response in Rasmussen encephalitis.
Journal of neuroinflammation
Owens GC,Erickson KL,Malone CC,Pan C,Huynh MN,Chang JW,Chirwa T,Vinters HV,Mathern GW,Kruse CA
MA1-21454 was used in flow cytometry to discuss the role of T cells in Rasmussen encephalitis
Sun Jul 19 00:00:00 EDT 2015
Cytokine diversity in the Th1-dominated human anti-influenza response caused by variable cytokine expression by Th1 cells, and a minor population of uncommitted IL-2+IFNγ- Thpp cells.
PloS one
Deng N,Weaver JM,Mosmann TR
MA1-21454 was used in flow cytometry to investigate the anti-influenza responses of T cell subsets
Thu Dec 18 00:00:00 EST 2014
Sickle cell vaso-occlusion causes activation of iNKT cells that is decreased by the adenosine A2A receptor agonist regadenoson.
Blood
Field JJ,Lin G,Okam MM,Majerus E,Keefer J,Onyekwere O,Ross A,Campigotto F,Neuberg D,Linden J,Nathan DG
MA1-21454 was used in flow cytometry to report results from a phase 1 trial testing the A2AR agonist regadenoson in adults with sickle cell disease.
Thu Apr 25 00:00:00 EDT 2013
Heterologous vaccination against human tuberculosis modulates antigen-specific CD4+ T-cell function.
European journal of immunology
Dintwe OB,Day CL,Smit E,Nemes E,Gray C,Tameris M,McShane H,Mahomed H,Hanekom WA,Scriba TJ
MA1-21454 was used in flow cytometry to characterize T cells after MVA85A vaccination
Sun Sep 01 00:00:00 EDT 2013
NF-κB is activated in CD4+ iNKT cells by sickle cell disease and mediates rapid induction of adenosine A2A receptors.
PloS one
Lin G,Field JJ,Yu JC,Ken R,Neuberg D,Nathan DG,Linden J
MA1-21454 was used in flow cytometry to study iNKT cells and A2ARs in sickle cell disease.
Fri Jul 11 00:00:00 EDT 2014
Disease-specific and inflammation-independent stromal alterations in spondylarthritis synovitis.
Arthritis and rheumatism
Yeremenko N,Noordenbos T,Cantaert T,van Tok M,van de Sande M,Cañete JD,Tak PP,Baeten D
MA1-21454 was used in immunohistochemistry to identify novel disease-specific pathways in spondylarthritis synovitis using geneome-wide microarray analysis
Tue Jan 01 00:00:00 EST 2013
Low levels of peripheral CD161++CD8+ mucosal associated invariant T (MAIT) cells are found in HIV and HIV/TB co-infection.
PloS one
Wong EB,Akilimali NA,Govender P,Sullivan ZA,Cosgrove C,Pillay M,Lewinsohn DM,Bishai WR,Walker BD,Ndung'u T,Klenerman P,Kasprowicz VO
MA1-21454 was used in flow cytometry to study CD161 high CD8+ T cells in an HIV and Mycobacterium tuberculosis endemic African setting.
Mon Aug 25 00:00:00 EDT 2014
Functional deficits of pertussis-specific CD4+ T cells in infants compared to adults following DTaP vaccination.
Clinical and experimental immunology
Sharma SK,Pichichero ME
MA1-21454 was used in flow cytometry to determine the function and phenotypes of antigen-specific CD4(+) T cells in adults compared to infants upon acellular pertussis vaccine administration
Sat Sep 01 00:00:00 EDT 2012
The acute environment, rather than T cell subset pre-commitment, regulates expression of the human T cell cytokine amphiregulin.
PloS one
Qi Y,Operario DJ,Georas SN,Mosmann TR
MA1-21454 was used in flow cytometry to analyze the regulation of Amphiregulin expression in human T cell subsets
Thu Dec 13 00:00:00 EST 2012
Quantitative tracking of T cell clones after haematopoietic stem cell transplantation.
EMBO molecular medicine
Mamedov IZ,Britanova OV,Bolotin DA,Chkalina AV,Staroverov DB,Zvyagin IV,Kotlobay AA,Turchaninova MA,Fedorenko DA,Novik AA,Sharonov GV,Lukyanov S,Chudakov DM,Lebedev YB
MA1-21454 was used in flow cytometry to track T cells after transplantation
Fri Apr 01 00:00:00 EDT 2011
B cell antigen receptor signaling enhances IFN-gamma-induced Stat1 target gene expression through calcium mobilization and activation of multiple serine kinase pathways.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research
Xu W,Nair JS,Malhotra A,Zhang JJ
MA1-21454 was used in flow cytometry to study the mechanism by which B cell antigen receptor signaling enhances IFN-gamma-induced Stat1 target gene expression
Tue Feb 01 00:00:00 EST 2005
Retention of unassembled components of integral membrane proteins by calnexin.
Science (New York, N.Y.)
Rajagopalan S,Xu Y,Brenner MB
MA1-21454 was used in immunoprecipitation to investigate calnexin's interaction with incompletely assembled TCR components
Fri Jan 21 00:00:00 EST 1994
Developmentally regulated rearrangement and expression of genes encoding the T cell receptor-T3 complex.
Cell
Furley AJ,Mizutani S,Weilbaecher K,Dhaliwal HS,Ford AM,Chan LC,Molgaard HV,Toyonaga B,Mak T,van den Elsen P
MA1-21454 was used in flow cytometry to review TCR alpha/beta-T3 complex expression and formation
Fri Jul 04 00:00:00 EDT 1986