Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 3
Positive Samples: HeLa, 293T, NIH/3T3, SH-SY5Y, Mouse lung, Rat brain
Immunogen sequence: WKPTFLSNEE FTQLMLEALD GFFLAIMTDG SIIYVSESVT SLLEHLPSDL VDQSIFNFIP EGEHSEVYKI LSTHLLESDS LTPEYLKSKN QLEFCCHMLR G
This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHe8; circadian locomoter output cycles kaput; circadian locomoter output cycles kaput protein; Circadian locomoter output cycles protein kaput; Class E basic helix-loop-helix protein 8; clock homolog; hCLOCK; mCLOCK; rCLOCK
Gene Aliases: 5330400M04Rik; BHLHE8; CLOCK; KAT13D; KIAA0334
UniProt ID: (Human) O15516, (Mouse) O08785, (Rat) Q9WVS9
Entrez Gene ID: (Human) 9575, (Mouse) 12753, (Rat) 60447
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support