Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The function of this protein is still unknown. The gene of human hypothetical protein (LOC54675) is up-regulated in colon cancer metastasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cardiolipin synthase (CMP-forming); CLS; Protein GCD10 homolog
Gene Aliases: 0610009I22Rik; 4930557M15Rik; 5730490M08Rik; C20orf155; CLS; CLS1; CRLS1; dJ967N21.6; GCD10
UniProt ID: (Human) Q9UJA2, (Mouse) Q80ZM8
Entrez Gene ID: (Human) 54675, (Mouse) 66586
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support