Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This gene encodes the beta subunit of casein kinase II, a ubiquitous protein kinase which regulates metabolic pathways, signal transduction, transcription, translation, and replication. The enzyme is composed of three subunits, alpha, alpha prime and beta, which form a tetrameric holoenzyme. The alpha and alpha prime subunits are catalytic, while the beta subunit serves regulatory functions. The enzyme localizes to the endoplasmic reticulum and the Golgi apparatus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alternative name: G5a, phosvitin; casein kinase 2, beta polypeptide; casein kinase 2, beta subunit; casein kinase II b subunit splicing isoform 132; casein kinase II b subunit splicing isoform 247; casein kinase II b subunit splicing isoform 318; casein kinase II b subunit splicing isoform 660; casein kinase II b subunit splicing isoform 806; Casein kinase II beta subunit; Casein kinase II subunit beta; chimera CSNK2B-LY6G5B splicing isoform 1072; chimera CSNK2B-LY6G5B splicing isoform 1103; chimera Csnk2b-Ly6g5b splicing isoform 2050; chimera Csnk2b-Ly6g5b splicing isoform 2275; chimera Csnk2b-Ly6g5b splicing isoform 2531; chimera CSNK2B-LY6G5B splicing isoform 532; chimera CSNK2B-LY6G5B splicing isoform 560; chimera CSNK2B-LY6G5B splicing isoform 562; chimera Csnk2b-Ly6g5b splicing isoform 901; CK II beta; Csnk2b splicing isoform 280; Csnk2b splicing isoform 662; Csnk2b-Ly6g5b; CSNK2B-LY6G5B-1072; CSNK2B-LY6G5B-1103; CSNK2B-LY6G5B-532; CSNK2B-LY6G5B-560; CSNK2B-LY6G5B-562; DADB-127H9.2; MGC138222; MGC138224; Phosvitin; Protein G5a
Gene Aliases: CK II beta; Ck2; CK2B; CK2N; CSK2B; CSNK2B; G5A
UniProt ID: (Human) P67870, (Rat) P67874, (Mouse) P67871
Entrez Gene ID: (Human) 1460, (Rat) 81650, (Mouse) 13001
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support