Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DNWSAAWTA ERNLNDLRKE NAHNRQKLTE IEFKIKLLEK DPYGLDVPNT AFGRQHSPYG PSPLGWPSSE TRASLYPPTL LEGPLRLSPL LPRGGGRGSR GPGNPPDHQI TKERGESSCD RLTDPHRAPS DAGPLAPPWE QDYRMMFPPP GQSYPDSALP PQRQDRFYSN CARLSGPAEL RSFNMPSLDK MDGSMPSEME SSRNDTKDNL GNLKVPDSSL PAENEATGPG FVPPPLAPIR GLLFPVDTRG PFIRRGPPFP PPPPGTVFGA SPDYFSPRDV PGPPRAPFAM RNVYLPRGFL PYRPPRPAFF PPAPTF (466-780 aa encoded by BC031065)
CTAGE1 (Cutaneous T Cell Lymphoma-Associated Antigen 1) is a Protein Coding gene. Diseases associated with CTAGE1 include Cutaneous T Cell Lymphoma and Lymphoma. An important paralog of this gene is MIA2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 21.2; cancer/testis antigen family 21, member 1; cancer/testis antigen family 21, member 2; CT21.2; cTAGE family member 2; cutaneous T-cell lymphoma-associated antigen 2; Protein cTAGE-2
Gene Aliases: CT21.1; CT21.2; CTAGE; CTAGE-1; CTAGE-2; CTAGE1; CTAGE2
UniProt ID: (Human) Q96RT6
Entrez Gene ID: (Human) 64693
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support