Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Immunogen Sequence: AQAVAVHSQD QSIPPCPQTW RSLWIGYSFL MHTGAGDQGG GQALMSPGSC LEDFRAAPFL ECQGRQGTCH FFANKYSFWL TTVKADLQFS SAPAPDTLKE SQAQRQKISR CQVCVKYS
Collagen IV is a type of collagen protein that is a major component of the basement membrane in various tissues, including the kidneys, lungs, and skin. It forms a network-like structure that provides support and stability to the surrounding tissue. Collagen IV is composed of three alpha chains, which are encoded by the COL4A1, COL4A2, and COL4A3 genes. Mutations in these genes can lead to various genetic disorders, such as Alport syndrome and Goodpasture syndrome, which affect the kidneys and lungs, respectively.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha 1 type IV collagen; alpha-2 collagen type IV; alpha1(IV) collagen; arresten; canstatin; COL4A1 NC1 domain; col4a3; col4a4; Collagen 4; Collagen alpha-1(IV) chain; Collagen alpha-2(IV) chain; Collagen alpha-3(IV) chain; Collagen alpha-4(IV) chain; Collagen alpha-5(IV) chain; Collagen alpha-6(IV) chain; collagen IV, alpha-1 polypeptide; collagen IV, alpha-3 polypeptide; Collagen IV, alpha-4 polypeptide; collagen IV, alpha-5 polypeptide; collagen IV, alpha-6 polypeptide; collagen of basement membrane, alpha-1 chain; collagen of basement membrane, alpha-4 chain; collagen of basement membrane, alpha-5 chain; collagen of basement membrane, alpha-6; collagen type IV alpha3 chain; collagen type IV alpha5 chain; collagen, type IV, alpha 1; collagen, type IV, alpha 3 (Goodpasture antigen); collagen, type IV, alpha 4; collagen, type IV, alpha 5; collagen, type IV, alpha 6; dA149D17.3; dA24A23.1; Del(8)Bru44H; dJ889N15.4 (Collagen Alpha 6(IV)); Goodpasture antigen; procollagen type IV alpha 5; procollagen type IV alpha 6; procollagen, type IV, alpha 1; procollagen, type IV, alpha 2; procollagen, type IV, alpha 3; procollagen, type IV, alpha 4; procollagen, type IV, alpha 5; procollagen, type IV, alpha 6; retinal anterior wiring; tumstatin; Type IV Collagen
Gene Aliases: [a]3(IV); [a]4(IV); alpha3(IV); ASLN; ATS; BB116301; Bru; BSVD; CA44; CA54; Col4a-1; Col4a-2; COL4A1; COL4A2; COL4A3; COL4A4; COL4A5; COL4A6; CXDELq22.3; Del(8)44H; DELXq22.3; DFNX6; E130010M05Rik; ICH; POREN2; RATOR; Raw; RGD1565499; Svc
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support