Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene is located within a chromosomal region that shows loss of heterozygosity in some bladder cancers. It contains a 5' CpG island that may be a frequent target of hypermethylation, and it may undergo hypermethylation-based silencing in some bladder cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA574M5.1 (deleted in bladder cancer chromosome region candidate 1 (IB3089A)); BMP/retinoic acid-inducible neural-specific protein (BRINP); BMP/retinoic acid-inducible neural-specific protein 1; BMP/retinoic acid-inducible neural-specific protein-1; bone morphogenetic protein/retinoic acid inducible neural-specific 1; bone morphogenic protein/retinoic acid inducible neural-specific 1; deleted in bladder cancer 1; deleted in bladder cancer chromosome region candidate 1; Deleted in bladder cancer protein 1; Deleted in bladder cancer protein 1 homolog
Gene Aliases: BRINP; BRINP1; DBC1; DBCCR1; FAM5A; IB3089A
UniProt ID: (Human) O60477, (Mouse) Q920P3
Entrez Gene ID: (Human) 1620, (Pig) 100625235, (Mouse) 56710
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support