Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MWGDLWLLP PASANPGTGT EAEFEKAAEE VRHLKTKPSD EEMLFIYGHY KQATVGDINT ERPGMLDFTG KAKWDAWNEL KGTSKEDAMK AYINKVEELK KKY (1-102 aa encoded by BC062996)
This gene encodes diazepam binding inhibitor, a protein that is regulated by hormones and is involved in lipid metabolism and the displacement of beta-carbolines and benzodiazepines, which modulate signal transduction at type A gamma-aminobutyric acid receptors located in brain synapses. The protein is conserved from yeast to mammals, with the most highly conserved domain consisting of seven contiguous residues that constitute the hydrophobic binding site for medium- and long-chain acyl-Coenzyme A esters. Diazepam binding inhibitor is also known to mediate the feedback regulation of pancreatic secretion and the postprandial release of cholecystokinin, in addition to its role as a mediator in corticotropin-dependent adrenal steroidogenesis. Three pseudogenes located on chromosomes 6, 8 and 16 have been identified. Multiple transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACBD 1; ACBP; acyl coenzyme A binding protein; acyl-CoA binding protein; Acyl-CoA-binding protein; acyl-Coenzyme A binding domain containing 1; CCK RP; CCKRP; cholecystokinin-releasing peptide, trypsin-sensitive; DBI; Diazepam binding inhibitor (GABA receptor modulator acyl-Coenxyme A binding protein); diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein); Diazepam binding inhibitor (GABA receptor modulator, acyl-Coenxyme A binding protein); diazepam binding inhibitor (GABA receptor modulator, acyl-Coenzyme A binding protein); diazepam binding inhibitor, splice form 1b; Diazepam-binding inhibitor; Endozepine; EP; GABA receptor modulator; LRRGT00046; MGC70414; octadecaneuropeptide; triakontatetraneuropeptide; valium
Gene Aliases: ACBD1; ACBP; Acoabp3; CCK-RP; DBI; endozepine; EP; Odn; RNACOABP3; Ttn
UniProt ID: (Human) P07108, (Mouse) P31786, (Rat) P11030
Entrez Gene ID: (Human) 1622, (Mouse) 13167, (Rat) 25045
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support