Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The RNA lariat debranching enzyme, or DBR1, specifically hydrolyzes 2-prime-to-5-prime branched phosphodiester bonds at the branch point of excised lariat intron RNA and converts them into linear molecules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: debranching enzyme homolog 1; Lariat debranching enzyme; RNA lariat debranching enzyme
Gene Aliases: AW018415; DBR1
UniProt ID: (Human) Q9UK59, (Mouse) Q923B1
Entrez Gene ID: (Human) 51163, (Pig) 106505697, (Rat) 681234, (Mouse) 83703
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support