Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Regulates the p53 pathway to control the expansion growth of digestive organs. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Digestive organ expansion factor homolog; digestive-organ expansion factor homolog; U3 small nucleolar RNA-associated protein 25 homolog; UTP25 small subunit processor component
Gene Aliases: AA408296; AW045375; C1orf107; DEF; DIEXF; DJ434O14.5; UTP25
UniProt ID: (Human) Q68CQ4, (Mouse) Q8BTT6
Entrez Gene ID: (Human) 27042, (Mouse) 215193
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support