Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MRGRRGDRMT INIQEHMAIN VCPGPIRPIR QISDYFPRGP GPEGGGGSGG EAPAHLVPLA LAPPAALLGA TTPEDGAEVD SYDSDDATAL GTLEFDLLYD RASCTLHCSI LRAKGLKPMD FNGLADPYVK LHLLPGACKA NKLKTKTQRN TLNPVWNEDL TYSGITDDDI THKVLRIAVC DEDKLSHNEF IGEIRVPLRR LKPSQKKHFN ICLERQVPLA SPSSMSAALR GISCYLKELE QAEQGQGLLE ERGRILLSLS YSSRRRGLLV GILRCAHLAA MDVNGYSDPY VKTYLRPDVD KKSKHKTCVK KKTLNPEFNE EFFYEIELST LATKTLEVTV WDYDIGKSND FIGGVSLGPG ARGEARKHWS DCLQQPDAAL ERWHTLTSEL PPAAGALSSA
There are at least two protein isoforms of the Double C2 protein, namely alpha (DOC2A) and beta (DOC2B), which contain two C2-like domains. DOC2A and DOC2B are encoded by different genes; these genes are at times confused with the unrelated DAB2 gene which was initially named DOC-2. DOC2A is mainly expressed in brain and is suggested to be involved in Ca(2+)-dependent neurotransmitter release.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Doc2; doc2-alpha; Double C2-like domain-containing protein alpha; double C2-like domains, alpha
Gene Aliases: Doc2; DOC2A
UniProt ID: (Human) Q14183
Entrez Gene ID: (Human) 8448
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support