Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total DPEP2.
Probable metalloprotease which hydrolyzes leukotriene D4 (LTD4) into leukotriene E4 (LTE4).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dipeptidase 2; MBD-2; Membrane-bound dipeptidase 2; putative membrane-bound dipeptidase 2
Gene Aliases: DPEP2; F630103D06Rik; MBD-2; MBD2; UNQ284/PRO323
UniProt ID: (Human) Q9H4A9, (Rat) Q5M872, (Mouse) Q8C255
Entrez Gene ID: (Human) 64174, (Rat) 291984, (Mouse) 319446
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support