Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EVCPEDRPL IVQFCANDPE VFVQAALLAQ DYCDAIDLNL GCPQMIAKRG HYGAFLQDEW DLLQRMILLA HEKLSVPVTC KIRVFPEIDK TVRYAQMLEK AGCQLLTVHG RTKEQKGPLS GAASWEHIKA VRKAVAIPVF ANGNIQCLQD VERCLRDTGV QGVMSAEGNL HNPALFEGRS PAVWELAEEY LDIVREHPCP LSYVRAHLFK LWHHTLQVHQ ELREELAKVK TLEGIAAVSQ ELKLRCQEEI SRQEGAKPTG DLPFHWICQP YIRPGPREGS KEKAGARSKR ALEEEEGGTE VLSKNKQKKQ LRNPHKTFDP SLKPKYAKCD QCGNPKGNRC VFSLCRGCCK KRASKETADC PGHGLLFKTK LEKSLAWKEA QPELQEPQPA APGTPGGFSE VMGSALA (1-406 aa encoded by BC017081)
Catalyzes the synthesis of dihydrouridine, a modified base found in the D-loop of most tRNAs. Target protein has a region of similarity to tRNA dihydrouridine synthase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dihydrouridine synthase 1-like; embryo-related protein; Liver regeneration-related protein LRRG08/LRRG09; tRNA-dihydrouridine synthase 1-like; tRNA-dihydrouridine(16/17) synthase [NAD(P)(+)]-like
Gene Aliases: 1110032N12Rik; DUS1; DUS1L; PP3111; x85
UniProt ID: (Human) Q6P1R4, (Rat) Q8K582, (Mouse) Q8C2P3
Entrez Gene ID: (Human) 64118, (Rat) 246185, (Mouse) 68730
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support