Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive Samples: Jurkat, SH-SY5Y, HeLa, Mouse testis, Mouse brain, Mouse skeletal muscle, Rat brain, Rat testis
Immunogen sequence: MSDKSDLKAE LERKKQRLAQ IREEKKRKEE ERKKKEADMQ QKKEPVQDDS DLDRKRRETE ALLQSIGISP EPPLVPTPMS PSSKSVSTPS EAGSQDSGDL GPLTRTLQWD TDPSVLQLQS DSELGRRLHK LGVSKVTQVD FLPREVVSYS KETQTPLATH QSEEDEEDEE MVESKVGQDS ELENQDKKQE VKEAPPRELT EEEKQQILHS EEFLIFFDRT
Acts as one of several non-catalytic accessory components of the cytoplasmic dynein 1 complex that are thought to be involved in linking dynein to cargos and to adapter proteins that regulate dynein function. Cytoplasmic dynein 1 acts as a motor for the intracellular retrograde motility of vesicles and organelles along microtubules. The intermediate chains mediate the binding of dynein to dynactin via its 150 kDa component (p150-glued) DCNT1. May play a role in mediating the interaction of cytoplasmic dynein with membranous organelles and kinetochores.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytoplasmic dynein 1 intermediate chain 1; Cytoplasmic dynein intermediate chain 1; cytoplasmic dynein intermediate chain 1 isoform 1.6; DH IC-1; Dnci 1; Dynein intermediate chain 1, cytosolic; Dynein intermediate chain 1, cytosolic (DH IC-1) (Cytoplasmic dynein intermediate chain 1); dynein, cytoplasmic 1, intermediate chain 1; dynein, cytoplasmic, intermediate chain 1; dynein, cytoplasmic, intermediate polypeptide 1
Gene Aliases: DH IC-1; DHIC-1; DIC; DNCI1; DNCIC1; DYNC1I1; IC74
UniProt ID: (Human) O14576, (Mouse) O88485, (Rat) Q63100
Entrez Gene ID: (Human) 1780, (Mouse) 13426, (Rat) 29564
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support