Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DYRK2 (also known as dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2) belongs to a family of protein kinases whose members are presumed to be involved in cellular growth and/or development and is defined by structural similarity of their kinase domains and their capability to autophosphorylate on tyrosine residues. DYRK2 expressed in E. coli demonstrated tyrosine autophosphorylation and catalyzed phosphorylation of histones H3 and H2B in vitro. DYRK2 also regulates p53 to induce apoptosis in response to DNA damage.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dual specificity tyrosine-(Y)-phosphorylation regulated kinase 2; Dual specificity tyrosine-phosphorylation-regulated kinase 2; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2; dyrk2
Gene Aliases: 1810038L18Rik; DYRK2
UniProt ID: (Human) Q92630, (Mouse) Q5U4C9
Entrez Gene ID: (Human) 8445, (Mouse) 69181, (Rat) 314862
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support