Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Positive test controls include: HepG2.
Immunogen sequence: MEFLWAPLLG LCCSLAAADR HTVFWNSSNP KFRNEDYTIH VQLNDYVDII CPHYEDHSVA DAAMEQYILY LVEHEEYQLC QPQSKDQVRW QCNRPSAKHG
This gene encodes a member of the ephrin family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B class, which are transmembrane proteins. This gene encodes an EFNA class ephrin which binds to the EPHA2, EPHA4, EPHA5, EPHA6, and EPHA7 receptors. Two transcript variants that encode different isoforms were identified through sequence analysis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EPH-related receptor tyrosine kinase ligand 1; Ephrin-A1; Immediate early response protein B61; LERK-1; ligand of eph-related kinase 1; TNF alpha-induced protein 4; Tumor necrosis factor alpha-induced protein 4; tumor necrosis factor, alpha-induced protein 4
Gene Aliases: AI325262; B61; ECKLG; EFL1; EFNA1; Epgl1; Epl1; EPLG1; LERK-1; LERK1; TNFAIP4
UniProt ID: (Human) P20827, (Mouse) P52793
Entrez Gene ID: (Human) 1942, (Mouse) 13636
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support