Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FAM129B (Family With Sequence Similarity 129 Member B, Niban Like Protein 1) is a Protein Coding gene. May play a role in apoptosis suppression. May promote melanoma cell invasion in vitro. Among its related pathways are MAP Kinase Signaling. An important paralog of this gene is FAM129A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 129, member B; Meg-3; Melanoma invasion by ERK; MINERVA; Niban-like protein 1; Protein FAM129B; Protein Niban 2
Gene Aliases: 9130404D14Rik; bA356B19.6; C9orf88; FAM129B; MEG-3; MINERVA; NIBAN2; OC58; RGD1307018
UniProt ID: (Human) Q96TA1, (Mouse) Q8R1F1, (Rat) B4F7E8
Entrez Gene ID: (Human) 64855, (Mouse) 227737, (Rat) 362115
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support