Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 150 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in mouse and rat.
Specificity of this antibody: FGF12.
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. This growth factor lacks the N-terminal signal sequence present in most of the FGF family members, but it contains clusters of basic residues that have been demonstrated to act as a nuclear localization signal. When transfected into mammalian cells, this protein accumulated in the nucleus, but was not secreted. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FGF; FGF-12; FHF-1; Fibroblast growth factor; Fibroblast growth factor 12; fibroblast growth factor 12B; fibroblast growth factor FGF-12b; Fibroblast growth factor homologous factor 1; fibroblast growth factor homologous factor 1b; Myocyte-activating factor
Gene Aliases: AV114868; B230343J05Rik; FGF-12; FGF12; FGF12B; FHF-1; FHF1; Fhf1b
UniProt ID: (Human) P61328, (Rat) P61150, (Mouse) P61329
Entrez Gene ID: (Human) 2257, (Rat) 170630, (Mouse) 14167
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support