Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAVYVGMLRL GRLCAGSSGV LGARAALSRS WQEARLQGVR FLSSREVDRM VSTPIGGLSY VQGCTKKHLN SKTVVQCLET TAQRVPEREA LVVLHEDVRL TFAQLKEEVD KAASGLLSIG LCKGDRLGMW GPNSYAWVLM QLATAQAGII LVSVNPAYQA MELEYVLKKV GCKALVFPKQ FKTQQYYNVL KQICPEVENA QPGALKSQRL PDLTTVISVD APLPGTLLLD EVVAAGSTRQ HLDQLQYNQQ FLSCHDPINI QFTSGTTGSP KGATLSHYNI VNNSNILGER LKLHEKTPEQ LRMILPNPLY HCLGSVAGTM MCLMYGATLI LASPIFNGKK ALEAISRERG TFLYGTPTMF VDILNQPDFS SYDISTMCGG VIAGSPAPPE LIRAIINKIN MKDLVVAYGT TENSPVTFAH FPEDTVEQKA ESVGRIMPHT EARIMNMEAG TLAKLNTPGE LCIRGYCVML GYWGEPQKTE EAVDQDKWYW TGDVATMNEQ GFCKIVGRSK DMIIRGGENI YPAELEDFFH THPKVQEVQV VGVKDDRMGE EICACIRLKD GEETTVEEIK AFCKGKISHF KIPKYIVFVT NYPLTISGKI QKFKLREQME RHLNL
Acyl-CoA synthases catalyze the initial reaction in fatty acid metabolism, by forming a thioester with CoA (PubMed:17762044). Has some preference toward medium-chain substrates (PubMed:17762044). Plays a role in adipocyte differentiation (PubMed:16380219). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acyl-CoA synthetase family member 2, mitochondrial; Medium-chain acyl-CoA ligase ACSF2, mitochondrial; PPARG binding, long chain fatty acid acyl Co-A ligase like
Gene Aliases: ACSF2; ACSMW; AVYV493; UNQ493/PRO1009
UniProt ID: (Human) Q96CM8, (Rat) Q499N5
Entrez Gene ID: (Human) 80221, (Rat) 619561
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support